Learn More
(S)-N-FMOC-α-Methylvaline, 98%, 98% ee, ACROS Organics™
Brand: Acros Organics 441151000
Code : 29
Additional Details : CAS Number : 169566-81-8 Weight : 0.05010kg Transport : UN number : 3175 Chem class : 1 Pack group : II
Quantity | 100mg |
---|
Chemical Identifiers
(S)-N-FMOC-α-Methylvaline | |
1634-04-4 | |
100.0 | |
98% | |
353.42 |
98% ee | |
97.5 | |
≥97.5% | |
C21H23NO4 | |
FMOC-3-Methyl-L-isovaline |
Specifications
(S)-N-FMOC-α-Methylvaline | |
100mg | |
1634-04-4 | |
97.5 | |
100.0 | |
+7.5° to +9.5° (20°C, 589nm) (c=1 in CHCl3) | |
≥97.5% | |
C21H23NO4 | |
FMOC-3-Methyl-L-isovaline |
White to pale yellow | |
98% ee | |
98.0% min. | |
Authentic | |
10.0% max. (MTBE) | |
Powder | |
98% | |
353.42 |