missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98%, 98% ee, ACROS Organics™

Brand:  Acros Organics 441091000

Code : 29

Additional Details : CAS Number : 1172127-44-4 Weight : 0.05010kg

Packaging Glass bottle

 View more versions of this product

Product Code. 11915561

  • £117.10 / 100mg


Estimated Shipment
20-03-2018
Add to basket

Specifications

Chemical Identifiers

(S)-N-FMOC-α-Methyl-2-fluorophenylalanine
1172127-44-4
100.0
98%
419.46
98% ee
97.5
98%
C25H22FNO4

Specifications

(S)-N-FMOC-α-Methyl-2-fluorophenylalanine
122.0° to 125.0°C
98% ee
98.0% min.
Authentic
Glass bottle
Powder
98%
419.46
White to pale yellow
100mg
1172127-44-4
97.5
100.0
− 47.00
98%
C25H22FNO4
Videos
SDS