missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-FMOC-Azetidine-2-carboxylic acid, 95%, 98% ee, ACROS Organics™

Brand:  Acros Organics 361600010

Code : 29

Additional Details : CAS Number : 136552-06-2 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10364212

  • £106.50 / 1g


Estimated Shipment
22-05-2018
Add to basket

Specifications

Chemical Identifiers

(S)-N-FMOC-Azetidine-2-carboxylic acid
136552-06-2
100.0
95%
323.35
(S)-N-(9-Fluorenylmethoxycarbonyl)-azetidine-2-carboxylic acid
95%, (98% e.e.)
94.0
≥94%
C19H17NO4
MFCD01320843

Specifications

(S)-N-FMOC-Azetidine-2-carboxylic acid
1g
136552-06-2
94.0
100.0
Powder
95%
323.35
(S)-N-(9-Fluorenylmethoxycarbonyl)-azetidine-2-carboxylic acid
White to light yellow
95%, (98% e.e.)
97.5% min. (HPLC)
Authentic
Glass bottle
≥94%
C19H17NO4
MFCD01320843
Videos
SDS