missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N-BOC-tert-Leucinol, 98%, Thermo Scientific Chemicals

Catalog Number p-4090114
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
153645-26-2
C11H23NO3
217.31
MFCD01861301
AZHJHZWFJVBGIL-MRVPVSSYSA-N
s---n-boc-tert-leucinol, n-boc-s-+-tert-leucinol, s-tert-butyl 1-hydroxy-3,3-dimethylbutan-2-yl carbamate, tert-butyl n-2s-1-hydroxy-3,3-dimethylbutan-2-yl carbamate, n-boc-l-tert-leucinol, s-n-boc-l-tert-leucinol, s-n-boc-tert-leucinol, s-2-tert-butoxycarbonylamino-3,3-dimethyl-1-butanol
10376058
tert-butyl N-[(2S)-1-hydroxy-3,3-dimethylbutan-2-yl]carbamate
CC(C)(C)C(CO)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
153645-26-2
C11H23NO3
217.31
MFCD01861301
AZHJHZWFJVBGIL-MRVPVSSYSA-N
s---n-boc-tert-leucinol, n-boc-s-+-tert-leucinol, s-tert-butyl 1-hydroxy-3,3-dimethylbutan-2-yl carbamate, tert-butyl n-2s-1-hydroxy-3,3-dimethylbutan-2-yl carbamate, n-boc-l-tert-leucinol, s-n-boc-l-tert-leucinol, s-n-boc-tert-leucinol, s-2-tert-butoxycarbonylamino-3,3-dimethyl-1-butanol
10376058
tert-butyl N-[(2S)-1-hydroxy-3,3-dimethylbutan-2-yl]carbamate
CC(C)(C)C(CO)NC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 104°C to 106°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 97.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation -5.5° to -7.9° (20°C, 589nm) (c=1, CHCl3)
Formula Weight 217.31
Percent Purity 98%
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-N-BOC-tert-Leucinol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'