missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-4-Fluorophenylglycine, 95%, 98% e.e., ACROS Organics™

Brand:  Acros Organics 361920010

Code : 29

Additional Details : CAS Number : 142186-36-5 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10572802

  • £254.80 / 1g


Estimated Shipment
14-02-2018
Add to basket

Specifications

Chemical Identifiers

(S)-N-BOC-4-Fluorophenylglycine
142186-36-5
100.0
95%
269.27
(L)-N-tert-Butoxycarbonyl-α-(4-fluorophenyl)acetic acid
95%, (98% e.e.)
94.0
95%
C13H16FNO4
MFCD01320862

Specifications

(S)-N-BOC-4-Fluorophenylglycine
142186-36-5
100.0
Authentic
Powder
95%
130.2°C
MFCD01320862
White
95%, (98% e.e.)
94.0
97.5% min. (HPLC)
Glass bottle
95%
C13H16FNO4
269.27
1g
(L)-N-tert-Butoxycarbonyl-α-(4-fluorophenyl)acetic acid
Videos
SDS