missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-(3-Pyridyl)alanine, 95%, 98% e.e., ACROS Organics™

Brand:  Acros Organics 361950010

Code : 29

Additional Details : CAS Number : 117142-26-4 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10048901

  • £92.90 / 1g


Estimated Shipment
21-05-2018
Add to basket

Specifications

Chemical Identifiers

(S)-N-BOC-(3-Pyridyl)alanine
117142-26-4
100.0
95%
266.29
N-tert-Butoxycarbonyl-3-pyridyl-L-alanine
95%, (98% e.e.)
94.0
≥94%
C13H18N2O4
MFCD00079681

Specifications

(S)-N-BOC-(3-Pyridyl)alanine
117142-26-4
100.0
Authentic
Crystalline Powder
95%
266.29
Off-white to pale yellow
1g
95%, (98% e.e.)
94.0
97.5% min. (HPLC)
Glass bottle
≥94%
C13H18N2O4
MFCD00079681
N-tert-Butoxycarbonyl-3-pyridyl-L-alanine
Videos
SDS