missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-2-Furylalanine tert-butylamine, 95%, 98% e.e., ACROS Organics™

Brand:  Acros Organics 366745000

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 881690-67-1 Weight : 0.05050kg

Quantity 500mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10048731

  • £91.70 / 500mg


Estimated Shipment
12-10-2017
Add to basket

Specifications

Chemical Identifiers

(S)-N-BOC-2-Furylalanine tert-butylamine
881690-67-1
100.0
C12H17NO5·C4H11N
MFCD04039764
95%, (98% e.e.)
94.0
95%
328.41

Specifications

(S)-N-BOC-2-Furylalanine tert-butylamine
881690-67-1
100.0
95%
C12H17NO5·C4H11N
MFCD04039764
95%, (98% e.e.)
94.0
Glass bottle
95%
328.41
500mg
Documentation

Technical Documents