missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-2-Bromophenylalanine, 95%, 98% e.e., ACROS Organics™

Brand:  Acros Organics 361675000

Code : 29

Additional Details : CAS Number : 261165-02-0 Weight : 0.05050kg

Quantity 500mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10088981

  • £53.70 / 500mg


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

(S)-N-BOC-2-Bromophenylalanine
261165-02-0
100.0
95%
344.21
N-tert-Butoxycarbonyl-2-bromophenyl-L-alanine
95%, (98% e.e.)
94.0
95%
C14H18BrNO4
MFCD01317709

Specifications

(S)-N-BOC-2-Bromophenylalanine
261165-02-0
100.0
Authentic
Crystalline Powder
95%
142.1°C
MFCD01317709
Pale yellow
95%, (98% e.e.)
94.0
97.5% min.
Glass bottle
95%
C14H18BrNO4
344.21
500mg
N-tert-Butoxycarbonyl-2-bromophenyl-L-alanine
Videos
SDS