missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N-Benzyl-α-methylbenzylamine, 98%, Thermo Scientific™

Catalog Number p-3777413
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17480-69-2
C15H24N
218.36
MFCD00066325
UHABCGJJMSQRRP-ZDUSSCGKSA-O
s---n-benzyl-1-phenylethylamine, s---n-benzyl-alpha-methylbenzylamine, s-n-benzyl-1-phenylethanamine, s-n-benzyl-1-phenylethylamine, benzyl 1s-1-phenylethyl amine, s-n-benzyl-alpha-methylbenzylamine, s--n-benzyl-alpha-phenylethylamine, pubchem5973, n-benzyl-d-amphetamine
1268085
C[C@H]([NH2+]CC1CCCCC1)C1=CC=CC=C1
This item is not returnable. View return policy
17480-69-2
C15H24N
218.36
MFCD00066325
UHABCGJJMSQRRP-ZDUSSCGKSA-O
s---n-benzyl-1-phenylethylamine, s---n-benzyl-alpha-methylbenzylamine, s-n-benzyl-1-phenylethanamine, s-n-benzyl-1-phenylethylamine, benzyl 1s-1-phenylethyl amine, s-n-benzyl-alpha-methylbenzylamine, s--n-benzyl-alpha-phenylethylamine, pubchem5973, n-benzyl-d-amphetamine
1268085
C[C@H]([NH2+]CC1CCCCC1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 129.0°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5620 to 1.5640
Quantity 1 g
Specific Gravity 1
Specific Rotation − 39.00
IUPAC Name (1S)-N-benzyl-1-phenylethanamine
Formula Weight 211.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-N-Benzyl-α-methylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-N-Benzyl-a-methylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'