missing translation for 'onlineSavingsMsg'
Learn More

(S)-alpha-Methylphenylalanine hydrate, 98%, 98% ee, Thermo Scientific™

Catalog Number 11905351
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
23239-35-2
C10H13NO2·xH2O
179.22
HYOWVAAEQCNGLE-JTQLQIEISA-N
alpha-methyl-l-phenylalanine, s-2-amino-2-methyl-3-phenylpropanoic acid, alpha-methyl-l-phe, alpha-methylphenylalanine, 2s-2-amino-2-methyl-3-phenylpropanoic acid, l-alpha-methylphenylalanine, alpha-benzyl-l-ala, s-2-amino-2-methyl-3-phenylpropionic acid, s-2-amimo-2-methyl-3-phenylpropanoic acid, s-alpha-methylphenylalanine
2724754
(2S)-2-amino-2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)(C(=O)O)N
This item is not returnable. View return policy
23239-35-2
C10H13NO2·xH2O
179.22
HYOWVAAEQCNGLE-JTQLQIEISA-N
alpha-methyl-l-phenylalanine, s-2-amino-2-methyl-3-phenylpropanoic acid, alpha-methyl-l-phe, alpha-methylphenylalanine, 2s-2-amino-2-methyl-3-phenylpropanoic acid, l-alpha-methylphenylalanine, alpha-benzyl-l-ala, s-2-amino-2-methyl-3-phenylpropionic acid, s-2-amimo-2-methyl-3-phenylpropanoic acid, s-alpha-methylphenylalanine
2724754
(2S)-2-amino-2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)(C(=O)O)N
CAS Min % 97.5
CAS Max % 100.0
Color White
Enantiomeric Excess (HPLC)> 99.0%
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 500 mg
Specific Rotation − 18.00
Formula Weight 179.22
Percent Purity 98%,98% ee
Physical Form Fine Crystalline Powder
Chemical Name or Material (S)-α-Methylphenylalanine hydrate, 99%, (99+% e.e.)
Show More Show Less
Product Identifier
  • (S)-alpha-Methylphenylalanine hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'