Learn More
Tocris Bioscience™ (S)-(+)-α-Methylhistamine dihydrobromide
Brand:  Tocris Bioscience™ 057250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13437773
  • £366.03 / 50mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Less active enantiomer of H3 agonist R-(-)-α-methylhistamine ; 120-fold less potent than R-(-) at H3.
R-enantiomer also available.
Specifications
Specifications
(S)-(+)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide
C6H11N3.2HBr
50mg
75614-93-6
Observed MW: 287
Soluble to 100 mM in water