Learn More
Tocris Bioscience™ (S)-(+)-α-Methylhistamine dihydrobromide
H3 agonist, less active enantiomer
POA
Specifications
Chemical Name or Material | (S)-(+)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide |
---|---|
CAS | 75614-93-6 |
Description
Less active enantiomer of H3 agonist R-(-)-α-methylhistamine ; 120-fold less potent than R-(-) at H3.R-enantiomer also available.
Specifications
(S)-(+)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide | |
75614-93-6 |
For Research Use Only