missing translation for 'onlineSavingsMsg'
Learn More

(S)-α-Methylaspartic acid, 98%, 98% e.e., ACROS Organics™

Brand:  Acros Organics 290135000

Code : 29

Additional Details : CAS Number : 3227-17-6 Weight : 0.05050kg

Quantity 500mg

 View more versions of this product

Product Code. 11955351

  • £99.60 / 500mg


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

(S)-α-Methylaspartic acid
3227-17-6
100.0
98%
HOOCC(CH3)NH2CH2COOH
(+)-2-Methyl-L-aspartic acid, (S)-(+)-2-Amino-2-methylbutanedioic acid
99%
97.5
98%
C5H9NO4
147.13

Specifications

(S)-α-Methylaspartic acid
99%
Authentic
100.0
+ 51.00
White
98%
98%
HOOCC(CH3)NH2CH2COOH
(+)-2-Methyl-L-aspartic acid, (S)-(+)-2-Amino-2-methylbutanedioic acid
98.0% min. (HPLC)
3227-17-6
97.5
Glass bottle
> 245.0°C
Fine Crystalline Powder or Powder
500mg
C5H9NO4
147.13
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (S)-alpha-Methylaspartic acid

Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.

SDS