missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 1.3400g/mL |
Boiling Point | 213.0°C to 214.0°C |
Flash Point | 89°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4680 to 1.4700 |
Show More |
Safety and Handling
Product Identifier
- (S)-(+)-alpha-Methoxy-alpha-trifluoromethylphenylacetyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'