Learn More
S-Isopropylisothiourea hydrobromide, Tocris Bioscience™
iNOS inhibitor, acts at arginine binding site
Brand:  Tocris (Bio-Techne) 089710
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13448123
  • £80.85 / 10mg
Estimated Shipment date
from Supplier 17-12-2019
Description
Description
Potent inhibitor of NO synthase, with selectivity for the inducible form.
Specifications
Chemical Identifiers
4269-97-0
199.11
s-isopropylisothiourea hydrobromide, s-isopropylthiourea hydrobromide, s-isopropyl-isothiourea hydrobromide, ipitu hydrobromide, d0j1lo, c4h10n2s.hbr, s-isopropylthiuronium bromide, s-isopropyl-itu hydrobromide, isopropyl carbamimidothioate hydrobromide
propan-2-yl carbamimidothioate;hydrobromide
C4H11BrN2S
SLGVZEOMLCTKRK-UHFFFAOYSA-N
9813062
CC(C)SC(=N)N.Br
Specifications
10mg
4269-97-0
s-isopropylisothiourea hydrobromide, s-isopropylthiourea hydrobromide, s-isopropyl-isothiourea hydrobromide, ipitu hydrobromide, d0j1lo, c4h10n2s.hbr, s-isopropylthiuronium bromide, s-isopropyl-itu hydrobromide, isopropyl carbamimidothioate hydrobromide
CC(C)SC(=N)N.Br
199.11
199.11
S-Isopropylisothiourea hydrobromide
C4H11BrN2S
SLGVZEOMLCTKRK-UHFFFAOYSA-N
propan-2-yl carbamimidothioate;hydrobromide
9813062
>98%