missing translation for 'onlineSavingsMsg'
Learn More

(S)-HexylHIBO, Tocris Bioscience™

Group I mGlu antagonist

Brand:  Tocris (Bio-Techne) 175050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13448703

  • £742.35 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Group I mGlu receptor antagonist (Kb values are 30 and 61 μM at mGlu1a and mGlu5a receptors respectively).

Racemate also available.

Specifications

Chemical Identifiers

334887-48-8
256.302
s-hexylhibo, s-hexylhomoibotenic acid, tocris-1749, tocris-1750, d0o0xx, ?s-?-amino-4-hexyl-2,3-dihydro-3-oxo-5-isoxazolepropanoic acid, 2s-2-amino-3-4-hexyl-3-oxo-1,2-oxazol-5-yl propanoic acid, s-3-3-hydroxy-4-hexylisoxazole-5-yl-2-aminopropanoic acid, 2s-2-amino-3-4-hexyl-3-oxo-2h-1,2-oxazol-5-yl propanoic acid, s-2-amino-3-4-hexyl-3-oxo-2,3-dihydroisoxazol-5-yl propanoic acid
(2S)-2-amino-3-(4-hexyl-3-oxo-1,2-oxazol-5-yl)propanoic acid
C12H20N2O4
OKJBLHIYOWSQDJ-VIFPVBQESA-N
6604908
CCCCCCC1=C(ONC1=O)CC(C(=O)O)N

Specifications

50mg
334887-48-8
s-hexylhibo, s-hexylhomoibotenic acid, tocris-1749, tocris-1750, d0o0xx, ?s-?-amino-4-hexyl-2,3-dihydro-3-oxo-5-isoxazolepropanoic acid, 2s-2-amino-3-4-hexyl-3-oxo-1,2-oxazol-5-yl propanoic acid, s-3-3-hydroxy-4-hexylisoxazole-5-yl-2-aminopropanoic acid, 2s-2-amino-3-4-hexyl-3-oxo-2h-1,2-oxazol-5-yl propanoic acid, s-2-amino-3-4-hexyl-3-oxo-2,3-dihydroisoxazol-5-yl propanoic acid
CCCCCCC1=C(ONC1=O)CC(C(=O)O)N
256.302
256.3
(S)-HexylHIBO
C12H20N2O4
OKJBLHIYOWSQDJ-VIFPVBQESA-N
(2S)-2-amino-3-(4-hexyl-3-oxo-1,2-oxazol-5-yl)propanoic acid
6604908
>99%
Videos
Documents