Learn More
Tocris Bioscience™ (S)-(-)-HA-966
Brand:  Tocris Bioscience™ 028250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13457523
  • £273.42 / 50mg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
Possesses potent sedative and ataxic action, probably through disruption of striatal dopaminergic mechanisms.
R-enantiomer also available.
Specifications
Specifications
(S)-(-)-3-Amino-1-hydroxypyrrolidin-2-one
>98%
Observed MW: 116.12
Soluble to 100 mM in water
111821-58-0
C4H8N2O2