missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Glycidyl nosylate, 98%, ACROS Organics™

Brand:  Acros Organics 296420100

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 115314-14-2 Weight : 0.06000kg


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10722194

  • £101.80 / 10g


EU Stock 1
Add to basket


Specifications Description & Specifications

Chemical Identifiers

(S)-(+)-Glycidyl nosylate
98%
259.23
≥97.5% (GC)
C9H9NO6S

Specifications

(S)-(+)-Glycidyl nosylate
Authentic
≥97.5% (GC)
Glass bottle
259.23
+ 21.50 (20.00°C c=2,CHCl3)
61° to 65°C
115314-14-2
Needles or Powder
98%
C9H9NO6S
(20°C, 589nm) (c=2, CHCl3) > +21.5°
Off-white to yellow-brown
10g

Safety and Handling

Product Identifier
  • (S)-(+)-Glycidyl nosylate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray
  • P280-Wear protective gloves/protective clothing/eye protection/face protection
  • P302+P352-IF ON SKIN: Wash with plenty of water/
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents