missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-Glycidol, 99+%, ee 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
Solubility
Soluble in chloroform, DMSO and methanol.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
Solubility
Soluble in chloroform, DMSO and methanol.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Color | Yellow |
Density | 1.116 g/mL |
Boiling Point | 66°C to 67°C (19 mmHg) |
Flash Point | 66°C (151°F) |
Refractive Index | 1.433 |
Quantity | 1 g |
UN Number | UN2810 |
Beilstein | 79783 |
Solubility Information | Soluble in chloroform,DMSO and methanol. |
Formula Weight | 74.08 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'