missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-α,α-Diphenyl-2-pyrrolidinemethanol, 99+%, Thermo Scientific Chemicals

Catalog Number p-122594
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
112068-01-6
C17H19NO
253.34
MFCD00075506
OGCGXUGBDJGFFY-INIZCTEOSA-N
s---alpha,alpha-diphenyl-2-pyrrolidinemethanol, s-diphenyl pyrrolidin-2-yl methanol, s---2-diphenylhydroxymethyl pyrrolidine, alpha,alpha-diphenyl-l-prolinol, diphenyl-l-pyrrolidin-2-yl-methanol, diphenyl-2s-pyrrolidin-2-yl methanol, s-alpha,alpha-diphenyl-2-pyrrolidinemethanol, s---diphenylpyrrolidinemethanol, s---alpha,alpha-diphenyl-2-pyrrolidine methanol, diphenyl 2s-pyrrolidin-2-ylmethanol
2724899
diphenyl-[(2S)-pyrrolidin-2-yl]methanol
C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
112068-01-6
C17H19NO
253.34
MFCD00075506
OGCGXUGBDJGFFY-INIZCTEOSA-N
s---alpha,alpha-diphenyl-2-pyrrolidinemethanol, s-diphenyl pyrrolidin-2-yl methanol, s---2-diphenylhydroxymethyl pyrrolidine, alpha,alpha-diphenyl-l-prolinol, diphenyl-l-pyrrolidin-2-yl-methanol, diphenyl-2s-pyrrolidin-2-yl methanol, s-alpha,alpha-diphenyl-2-pyrrolidinemethanol, s---diphenylpyrrolidinemethanol, s---alpha,alpha-diphenyl-2-pyrrolidine methanol, diphenyl 2s-pyrrolidin-2-ylmethanol
2724899
diphenyl-[(2S)-pyrrolidin-2-yl]methanol
C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 77°C to 80°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 1 g
Fieser 16,254
Specific Rotation Condition −59° (25°C c=3,methanol)
Specific Rotation -59
Formula Weight 253.34
Percent Purity 99+%
Chemical Name or Material (S)-(-)-α, α-Diphenyl-2-pyrrolidinemethanol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-a,a-Diphenyl-2-pyrrolidinemethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'