Learn More
Alfa Aesar™ [(S)-Binap RuCl p-cymene]Cl
Brand:  Alfa Aesar™ H36851.ME
UNSPSC : 12352100
Code : NEW
Additional Details : CAS Number : 130004-33-0 Weight : 0.00050kg
Quantity 500mg
 View more versions of this product
Product Code. 15427707
  • £66.89 / 500mg
Estimated Shipment date
from Supplier 11-08-2017
Add to basket
Specifications Description & Specifications
Chemical Identifiers
[(S)-Binap RuCl p-cymene]Cl
130004-33-0
928.87
MFCD00134456
Specifications
[(S)-Binap RuCl p-cymene]Cl
Crystalline Powder
MFCD00134456
500mg
130004-33-0
928.87
Orange
Air Sensitive
Documentation
Technical Documents