missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Bay K 8644, Tocris Bioscience™

CaV1.x activator; (S)-(-)-enantiomer of (±)-Bay K 8644

Brand:  Tocris (Bio-Techne) 1546/10

Code : Z1

Additional Details : Weight : 0.60000kg

Quantity 10mg

 View more versions of this product

Product Code. 13429653

  • £282.24 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

L-type Ca2+-channel activator with positive inotropic, vasoconstrictive and behavioral effects in vivo.

Racemate and (R)-(+)-Enantiomer also available.

Specifications

Chemical Identifiers

98625-26-4
356.301
s---bay k 8644, bay-k-8644 s---, --s-bayk8644, s---bay k8644, bay-k-8644 s--, methyl 4s-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate, s---1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester, tocris-1544, lopac-b-112, d02rih
methyl (4S)-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
C16H15F3N2O4
ZFLWDHHVRRZMEI-ZDUSSCGKSA-N
6603728
CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC

Specifications

10mg
98625-26-4
s---bay k 8644, bay-k-8644 s---, --s-bayk8644, s---bay k8644, bay-k-8644 s--, methyl 4s-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate, s---1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester, tocris-1544, lopac-b-112, d02rih
CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC
356.301
356.3
(S)-(-)-Bay K 8644
C16H15F3N2O4
ZFLWDHHVRRZMEI-ZDUSSCGKSA-N
methyl (4S)-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
6603728
>99%
Videos
Documents