Learn More
(S)-(-)-Atenolol, Tocris Bioscience™
β1-antagonist. Active isomer of atenolol (Cat. No. 0387)
Brand:  Tocris (Bio-Techne) 039310
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13447613
  • £116.13 / 10mg
Estimated Shipment date
from Supplier 23-03-2020
Description
Description
The active enantiomer of (RS)-atenolol (Cat. No. 0387), a cardioselective β-adrenergic blocker.
Specifications
Chemical Identifiers
93379-54-5
266.341
esatenolol, s-atenolol, --atenolol, s---atenolol, s--atenolol, esatenolol inn, unii-dpf757bosr, s---4-2-hydroxy-3-isopropylaminopropoxy phenylacetamide
CHEBI:31556
CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O
C14H22N2O3
METKIMKYRPQLGS-LBPRGKRZSA-N
175540
2-[4-[(2S)-2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
Specifications
10mg
93379-54-5
esatenolol, s-atenolol, --atenolol, s---atenolol, s--atenolol, esatenolol inn, unii-dpf757bosr, s---4-2-hydroxy-3-isopropylaminopropoxy phenylacetamide
CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O
266.341
CHEBI:31556
>97%
(S)-(-)-Atenolol
C14H22N2O3
METKIMKYRPQLGS-LBPRGKRZSA-N
2-[4-[(2S)-2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
175540
266.34