missing translation for 'onlineSavingsMsg'
Learn More

(S)-AMPA, Tocris Bioscience™

Selective AMPA agonist; active isomer of (RS)-AMPA (Cat. No. 0169)

Brand:  Tocris (Bio-Techne) 0254/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13447513

  • £943.74 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Active enantiomer of AMPA (EC50 = 3.5 μM).

NPEC-caged-(S)-AMPA, Inactive Enantiomer and Racemate also available.

Specifications

Chemical Identifiers

83643-88-3
186.167
s-ampa, s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, ampa, l, unii-9280sc28gd, s-2-amino-3-3-hydroxy-5-methyl-isoxazol-4-yl-propionic acid, amq, glutamate-ampa, 1ftm, tocris-0169, tocris-0254
(2S)-2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
C7H10N2O4
UUDAMDVQRQNNHZ-YFKPBYRVSA-N
158397
CC1=C(C(=O)NO1)CC(C(=O)O)N

Specifications

50mg
83643-88-3
s-ampa, s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, ampa, l, unii-9280sc28gd, s-2-amino-3-3-hydroxy-5-methyl-isoxazol-4-yl-propionic acid, amq, glutamate-ampa, 1ftm, tocris-0169, tocris-0254
CC1=C(C(=O)NO1)CC(C(=O)O)N
186.167
186.17
(S)-AMPA
C7H10N2O4
UUDAMDVQRQNNHZ-YFKPBYRVSA-N
(2S)-2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
158397
>99%
Videos
Documents