missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-α-Amino-γ-butyrolactone hydrobromide, 99%, Thermo Scientific Chemicals

Catalog Number 11936191
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
15295-77-9
C4H8BrNO2
182.02
MFCD00012723,MFCD00674493
MKLNTBLOABOJFZ-UHFFFAOYNA-N
s-3-aminodihydrofuran-2 3h-one hydrobromide, s---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s-2-amino-4-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, s---alpha-amino-gamma-butyrolactonehydrobromide, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr
16213138
Br.NC1CCOC1=O
15295-77-9
C4H8BrNO2
182.02
MFCD00012723,MFCD00674493
MKLNTBLOABOJFZ-UHFFFAOYNA-N
s-3-aminodihydrofuran-2 3h-one hydrobromide, s---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s-2-amino-4-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, s---alpha-amino-gamma-butyrolactonehydrobromide, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr
16213138
Br.NC1CCOC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 236.0°C to 238.9°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5 % min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition − 21.50 (20.00°C c=4,water)
Specific Rotation − 21.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone
IUPAC Name 3-aminooxolan-2-one hydrobromide
Formula Weight 182.02
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(-)-α-Amino-γ-butyrolactone hydrobromide
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-a-Amino-y-butyrolactone hydrobromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'