missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-2689289
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
17257-71-5
C10H8F3O3
233.17
MFCD00064200
JJYKJUXBWFATTE-VIFPVBQESA-M
s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
6992788
CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F
This item is not returnable. View return policy
17257-71-5
C10H8F3O3
233.17
MFCD00064200
JJYKJUXBWFATTE-VIFPVBQESA-M
s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
6992788
CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless or White
Density 1.3030g/mL
Boiling Point 116°C to 118°C (1.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C6H5C(OCH3)(CF3)CO2H
Refractive Index 1.4720 to 1.475
Quantity 250 mg
Fieser 11,329
Merck Index 15, 6365
Specific Gravity 1.303
Specific Rotation Condition - 71.00 (20.00°C c=1.6,CH3OH)
Specific Rotation -71
IUPAC Name (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
Formula Weight 234.17
Percent Purity 98%
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material (S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-a-Methoxy-a-(trifluoromethyl)phenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'