missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-Allylglycine, 98%, 98% ee, Thermo Scientific Chemicals

Catalog Number p-57667
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16338-48-0
C5H9NO2
115.13
MFCD00002627
WNNNWFKQCKFSDK-BYPYZUCNSA-N
l-allylglycine, s-2-aminopent-4-enoic acid, 2s-2-aminopent-4-enoic acid, h-gly ally-oh, s---2-amino-4-pentenoic acid, s-2-amino-4-pentenoic acid, s-allylglycine, l-2-amino-4-pentenoic acid, 3-vinyl-l-alanine
167529
(2S)-2-aminopent-4-enoic acid
C=CCC(C(=O)O)N
This item is not returnable. View return policy
16338-48-0
C5H9NO2
115.13
MFCD00002627
WNNNWFKQCKFSDK-BYPYZUCNSA-N
l-allylglycine, s-2-aminopent-4-enoic acid, 2s-2-aminopent-4-enoic acid, h-gly ally-oh, s---2-amino-4-pentenoic acid, s-2-amino-4-pentenoic acid, s-allylglycine, l-2-amino-4-pentenoic acid, 3-vinyl-l-alanine
167529
(2S)-2-aminopent-4-enoic acid
C=CCC(C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 253.0°C to 255.0°C
Color White
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2C=CHCH2CH(NH2)CO2H
Quantity 250 mg
Beilstein 04, 2852
Specific Rotation − 37.00
Solubility Information Solubility in water: soluble.
Formula Weight 115.13
Percent Purity 98%,98% ee
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-α-Allylglycine, 98%, (98% e.e.)
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-2-Amino-4-pentenoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'