missing translation for 'onlineSavingsMsg'
Learn More

S-Acetamidomethyl-L-cysteine hydrochloride, 98%, Thermo Scientific™

Catalog Number p-7054767
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
28798-28-9
C6H13ClN2O3S
228.691
MFCD00077080
SZWPOAKLKGUXDD-JEDNCBNOSA-N
h-cys acm-oh hcl, s-acetamidomethyl-l-cysteine hydrochloride, h-cys acm-oh.hcl, 2r-2-amino-3-acetamidomethyl sulfanyl propanoic acid hydrochloride, l-cysteine, s-acetylamino methyl-, monohydrochloride, h-cys acm .hcl, h-cys acm-oh hydrochloride, s-acetamidomethylcysteine hydrochloride, s-acetyl-aminomethyl-l-cysteine hydrochloride, h-cys acm-oh inverted exclamation mark currencyhcl
16211002
(2R)-3-(acetamidomethylsulfanyl)-2-aminopropanoic acid;hydrochloride
CC(=O)NCSCC(C(=O)O)N.Cl
This item is not returnable. View return policy
28798-28-9
C6H13ClN2O3S
228.691
MFCD00077080
SZWPOAKLKGUXDD-JEDNCBNOSA-N
h-cys acm-oh hcl, s-acetamidomethyl-l-cysteine hydrochloride, h-cys acm-oh.hcl, 2r-2-amino-3-acetamidomethyl sulfanyl propanoic acid hydrochloride, l-cysteine, s-acetylamino methyl-, monohydrochloride, h-cys acm .hcl, h-cys acm-oh hydrochloride, s-acetamidomethylcysteine hydrochloride, s-acetyl-aminomethyl-l-cysteine hydrochloride, h-cys acm-oh inverted exclamation mark currencyhcl
16211002
(2R)-3-(acetamidomethylsulfanyl)-2-aminopropanoic acid;hydrochloride
CC(=O)NCSCC(C(=O)O)N.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 165°C (decomposition)
Beilstein 4015821
Formula Weight 228.7
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material S-Acetamidomethyl-L-cysteine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'