missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone, 97%, Thermo Scientific Chemicals

Catalog Number p-3758200
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17342-08-4
C5H9NO2
115.13
MFCD00077792
HOBJEFOCIRXQKH-BYPYZUCNSA-N
l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847
643511
(5S)-5-(hydroxymethyl)pyrrolidin-2-one
C1CC(=O)NC1CO
This item is not returnable. View return policy
17342-08-4
C5H9NO2
115.13
MFCD00077792
HOBJEFOCIRXQKH-BYPYZUCNSA-N
l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847
643511
(5S)-5-(hydroxymethyl)pyrrolidin-2-one
C1CC(=O)NC1CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 78°C to 86°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition +30° (20°C c=2,C2H5OH)
Specific Rotation + 30
Formula Weight 115.13
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(+)-5-Hydroxymethyl-2-pyrrolidinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'