missing translation for 'onlineSavingsMsg'
Learn More

S-(4-Methoxybenzyl)-L-cysteine, tech. 80%, Thermo Scientific™

Catalog Number 15480849
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
2544-31-2
C11H15NO3S
241.305
MFCD00037207
PQPZSPJVMUCVAQ-JTQLQIEISA-N
h-cys pmeobzl-oh, h-cys 4-meo-bzl-oh, s-4-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxybenzyl thio propanoic acid, s-4-methoxybenzyl cysteine, s-p-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxy-benzylsulfanyl-propionic acid, 2-amino-3-4-methoxybenzyl thio propanoic acid, 2r-2-amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, ambotzhaa6100
7009563
(2R)-2-amino-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
COC1=CC=C(C=C1)CSCC(C(=O)O)N
This item is not returnable. View return policy
2544-31-2
C11H15NO3S
241.305
MFCD00037207
PQPZSPJVMUCVAQ-JTQLQIEISA-N
h-cys pmeobzl-oh, h-cys 4-meo-bzl-oh, s-4-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxybenzyl thio propanoic acid, s-4-methoxybenzyl cysteine, s-p-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxy-benzylsulfanyl-propionic acid, 2-amino-3-4-methoxybenzyl thio propanoic acid, 2r-2-amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, ambotzhaa6100
7009563
(2R)-2-amino-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
COC1=CC=C(C=C1)CSCC(C(=O)O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 241.31
Percent Purity 80%
Grade Technical
Quantity 5 g
Physical Form Solid
Chemical Name or Material S-(4-Methoxybenzyl)-L-cysteine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'