Learn More
(S)-4-Carboxy-3-hydroxyphenylglycine, Tocris Bioscience™
Group I mGlu antagonist; also group II mGlu agonist
Brand:  Tocris (Bio-Techne) 032050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13467543
  • £714.42 / 50mg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Competitive antagonist at group I mGlu1a/1a receptors, mixed effect at mGlu5a/5b receptors; agonist at group II metabotropic glutamate receptors.
Racemate also available.
Specifications
Chemical Identifiers
85148-82-9
211.173
s-4c3hpg, s-4-carboxy-3-hydroxyphenylglycine, dfs3siu1me, unii-dfs3siu1me, chembl39372, s-4c3h-pg, 4-carboxy-3-hydroxyphenylglycine, s, s-4-amino carboxy methyl-2-hydroxybenzoic acid, 4-1s-1-amino-2-hydroxy-2-oxoethyl-2-hydroxybenzoic acid, benzeneacetic acid, alpha-amino-4-carboxy-3-hydroxy-, alphas
4-[(S)-amino(carboxy)methyl]-2-hydroxybenzoic acid
C9H9NO5
GXZSAQLJWLCLOX-ZETCQYMHSA-N
5311455
C1=CC(=C(C=C1C(C(=O)O)N)O)C(=O)O
Specifications
(S)-4-Carboxy-3-hydroxyphenylglycine
C9H9NO5
GXZSAQLJWLCLOX-ZETCQYMHSA-N
4-[(S)-amino(carboxy)methyl]-2-hydroxybenzoic acid
5311455
>98%
85148-82-9
s-4c3hpg, s-4-carboxy-3-hydroxyphenylglycine, dfs3siu1me, unii-dfs3siu1me, chembl39372, s-4c3h-pg, 4-carboxy-3-hydroxyphenylglycine, s, s-4-amino carboxy methyl-2-hydroxybenzoic acid, 4-1s-1-amino-2-hydroxy-2-oxoethyl-2-hydroxybenzoic acid, benzeneacetic acid, alpha-amino-4-carboxy-3-hydroxy-, alphas
C1=CC(=C(C=C1C(C(=O)O)N)O)C(=O)O
211.173
211.17