missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-4-Boc-morpholine-2-carboxylic acid, 95%

Brand:  Alfa Aesar™ H64807.03

Code : NEW

Additional Details : CAS Number : 868689-63-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15422179

  • £450.50 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

(S)-4-Boc-morpholine-2-carboxylic acid
231.25
14461235
868689-63-8
MFCD06200662
(S)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid

Specifications

(S)-4-Boc-morpholine-2-carboxylic acid
Solid
231.25
14461235
127° to 133°C
868689-63-8
95%
MFCD06200662
(S)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid
1g
Videos
SDS