missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Benzyl-2-oxazolidinone, 99%, >99% ee, Thermo Scientific Chemicals

Catalog Number 11946831
Click to view available options
Quantity:
2.5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
90719-32-7
C10H11NO2
177.20
MFCD00064496
OJOFMLDBXPDXLQ-VIFPVBQESA-N
s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
736225
(4S)-4-benzyl-1,3-oxazolidin-2-one
O=C1N[C@@H](CC2=CC=CC=C2)CO1
This item is not returnable. View return policy
90719-32-7
C10H11NO2
177.20
MFCD00064496
OJOFMLDBXPDXLQ-VIFPVBQESA-N
s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
736225
(4S)-4-benzyl-1,3-oxazolidin-2-one
O=C1N[C@@H](CC2=CC=CC=C2)CO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 87.0°C to 90.0°C
Color White to Yellow
Assay Percent Range 98.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: insoluble
Specific Rotation − 62.00
Formula Weight 177.2
Percent Purity 99%,>99% ee
Specific Rotation Condition − 62.00 (20.00°C C=1,CHCl3)
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material (S)-4-Benzyl-2-oxazolidinone
Show More Show Less
compliance-icons
Product Identifier
  • (S)-4-Benzyl-2-oxazolidinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'