missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Hydroxyphenylglycine, Tocris Bioscience™

Group I mGlu agonist, active isomer

Brand:  Tocris (Bio-Techne) 032650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13467553

  • £563.01 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Agonist at group I metabotropic glutamate receptors (mGlu1) having no effect at mGlu2 or mGlu4.

Racemate also available.

Specifications

Chemical Identifiers

71301-82-1
167.164
s-3-hydroxyphenylglycine, 2s-2-amino-2-3-hydroxyphenyl acetic acid, s-2-3-hydroxyphenyl glycine, s-3hpg, s-3h-pg, s-amino-3-hydroxy-phenyl-acetic acid, s-2-amino-2-3-hydroxyphenyl acetic acid, 3hpg, tocris-0324, tocris-0326
(2S)-2-amino-2-(3-hydroxyphenyl)acetic acid
C8H9NO3
DQLYTFPAEVJTFM-ZETCQYMHSA-N
6604712
C1=CC(=CC(=C1)O)C(C(=O)O)N

Specifications

(S)-3-Hydroxyphenylglycine
C8H9NO3
DQLYTFPAEVJTFM-ZETCQYMHSA-N
(2S)-2-amino-2-(3-hydroxyphenyl)acetic acid
6604712
>99%
71301-82-1
s-3-hydroxyphenylglycine, 2s-2-amino-2-3-hydroxyphenyl acetic acid, s-2-3-hydroxyphenyl glycine, s-3hpg, s-3h-pg, s-amino-3-hydroxy-phenyl-acetic acid, s-2-amino-2-3-hydroxyphenyl acetic acid, 3hpg, tocris-0324, tocris-0326
C1=CC(=CC(=C1)O)C(C(=O)O)N
167.164
167.16
50mg
Videos
Documents