missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, ACROS Organics™

Brand:  Acros Organics 412120050

Code : 29

Additional Details : CAS Number : 36567-72-3 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10561714

  • £323.80 / 5g


Estimated Shipment
14-02-2018
Add to basket

Specifications

Chemical Identifiers

(S)-3-Hydroxy-3-phenylpropanoic acid
98.5
99%
C9H10O3
MFCD00145262
36567-72-3
100.0
99%
166.18

Specifications

(S)-3-Hydroxy-3-phenylpropanoic acid
White to almost white
5g
98.5
Authentic
Needle-Like Crystals
99%
C9H10O3
MFCD00145262
−22.50° (20°C c=4,CH3OH)
116.0° to 119.0°C
36567-72-3
100.0
Glass bottle
99%
− 22.50
166.18
Videos
SDS