missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane, 98%, Thermo Scientific™
Click to view available options
1g; Glass bottle
Specifications
CAS | 60456-22-6 |
Density | 1.0950g/mL |
Boiling Point | 63.0°C (37.0 mmHg) |
Flash Point | 50°C |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass Bottle |
Molecular Formula | C6H11ClO2 |
Refractive Index | 1.4330 to 1.4350 |
MDL Number | MFCD00273365 |
Quantity | 1g |
Show More |
Safety and Handling
Product Identifier
- (S)-(-)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H301-Toxic if swallowed.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'