missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Butyn-2-ol, 99%, Thermo Scientific Chemicals

Catalog Number p-7048153
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2914-69-4
C4H6O
70.091
MFCD00190166
GKPOMITUDGXOSB-BYPYZUCNSA-N
s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc
6995470
(2S)-but-3-yn-2-ol
CC(C#C)O
This item is not returnable. View return policy
2914-69-4
C4H6O
70.091
MFCD00190166
GKPOMITUDGXOSB-BYPYZUCNSA-N
s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc
6995470
(2S)-but-3-yn-2-ol
CC(C#C)O

(S)-(-)-3-Butyn-2-ol is used to prepare (S)-(-)-2-tert-butyldimethylsiloxybut-3-yne. It finds application in ruthenium-catalyzed polymerization to get helical polyacetylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Butyn-2-ol is used to prepare (S)-(-)-2-tert-butyldimethylsiloxybut-3-yne. It finds application in ruthenium-catalyzed polymerization to get helical polyacetylenes.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.891
Boiling Point 108°C to 109°C
Flash Point 28°C (82°F)
Refractive Index 1.426
Quantity 250 mg
UN Number UN2929
Beilstein 4652597
Solubility Information Miscible with water.
Optical Rotation −47° (Neat)
Formula Weight 70.09
Percent Purity 99%
Chemical Name or Material (S)-(-)-3-Butyn-2-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'