missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(BOC-Amino)piperidine, 97%, Thermo Scientific Chemicals

Catalog Number 10704122
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
216854-23-8
C10H20N2O2
200.28
WUOQXNWMYLFAHT-QMMMGPOBSA-N
s-3-boc-aminopiperidine, s-3-n-boc-aminopiperidine, s-tert-butyl piperidin-3-ylcarbamate, s-3-boc-amino piperidine, tert-butyl n-3s-piperidin-3-yl carbamate, s-3-tert-butoxycarbonylamino piperidine, tert-butyl s-piperidin-3-ylcarbamate, s-3-n-boc-amino piperidine, s---3-tert-butoxycarbonylaminopiperidine, 3s-3-aminopiperidine, 3-boc protected
1514171
tert-butyl N-[(3S)-piperidin-3-yl]carbamate
CC(C)(C)OC(=O)NC1CCCNC1
This item is not returnable. View return policy
216854-23-8
C10H20N2O2
200.28
WUOQXNWMYLFAHT-QMMMGPOBSA-N
s-3-boc-aminopiperidine, s-3-n-boc-aminopiperidine, s-tert-butyl piperidin-3-ylcarbamate, s-3-boc-amino piperidine, tert-butyl n-3s-piperidin-3-yl carbamate, s-3-tert-butoxycarbonylamino piperidine, tert-butyl s-piperidin-3-ylcarbamate, s-3-n-boc-amino piperidine, s---3-tert-butoxycarbonylaminopiperidine, 3s-3-aminopiperidine, 3-boc protected
1514171
tert-butyl N-[(3S)-piperidin-3-yl]carbamate
CC(C)(C)OC(=O)NC1CCCNC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 104.4°C to 104.5°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Assay Percent Range 96% min. (GC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 200.28
Percent Purity 97%
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-(BOC-Amino)piperidine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-3-(Boc-Amino)piperidine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'