missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-3-Amino-1-Boc-piperidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.
Solubility
Soluble in dimethylsulfoxide.
Notes
Air and carbon dioxide sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.
Solubility
Soluble in dimethylsulfoxide.
Notes
Air and carbon dioxide sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Specifications
Quantity | 250 mg |
UN Number | UN2735 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in dimethylsulfoxide. |
Formula Weight | 200.28 |
Percent Purity | 97% |
Chemical Name or Material | (S)-(+)-3-Amino-1-Boc-piperidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'