missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-1,2-propanediol, 98%, Thermo Scientific™

Catalog Number 15109303
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
61278-21-5
91.11
KQIGMPWTAHJUMN-VKHMYHEASA-N
s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol
440902
CHEBI:1456
(2S)-3-aminopropane-1,2-diol
C(C(CO)O)N
This item is not returnable. View return policy
61278-21-5
91.11
KQIGMPWTAHJUMN-VKHMYHEASA-N
s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol
440902
CHEBI:1456
(2S)-3-aminopropane-1,2-diol
C(C(CO)O)N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 53°C to 57°C
Color Colorless to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C3H9NO2
Quantity 5 g
Specific Rotation −27° to −31° (20°C, 589nm) (c=2, 1M HCl)
Formula Weight 91.11
Percent Purity ≥97.5% (Total base)
Physical Form Liquid to Low Melting Solid
Chemical Name or Material (S)-3-Amino-1, 2-propanediol
Show More Show Less
compliance-icons
Product Identifier
  • (S)-3-Amino-1,2-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'