missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals
63.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Quantity | 1 g |
Beilstein | 9161065 |
Formula Weight | 410.42 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | (S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'