Learn More
(S)-3,5-DHPG, Tocris Bioscience™
Selective group I mGlu agonist. Active enantiomer of 3,5-DHPG (Cat. No. 0342)
Brand:  Tocris (Bio-Techne) 0805/5
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 5mg
 View more versions of this product
Product Code. 13488023
  • £132.00 / 5mg
Estimated Shipment
07-03-2018
Add to basket
Description
Description
Selective group I mGlu receptor agonist. Also available as part of the Group I mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . (RS)-3,5-DHPG (Cat. No. 0342) also available.
Specifications
Chemical Identifiers
(S)-3, 5-DHPG
C8H9NO4
183.16
(S)-3, 5-Dihydroxyphenylglycine
Specifications
(S)-3, 5-DHPG
>98%
183.16
5mg
162870-29-3
C8H9NO4
(S)-3, 5-Dihydroxyphenylglycine
Videos