missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol, 96%

Brand:  Alfa Aesar™ H55445.MD

Code : NEW

Additional Details : CAS Number : 119707-74-3 Weight : 0.00025kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15451688

  • £66.36 / 250mg


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol
444.12
UN3077
119707-74-3
MFCD03093629

Specifications

(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol
Powder
444.12
UN3077
256° to 260°C
119707-74-3
96%
MFCD03093629
White to brown
250mg
Videos
SDS