missing translation for 'onlineSavingsMsg'
Learn More

S 25585, Tocris Bioscience™

Potent and selective NPY Y5 receptor antagonist

Brand:  Tocris (Bio-Techne) 343250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13453004

  • £570.36 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent neuropeptide Y (NPY) Y5 receptor antagonist (IC50 values are 5.4, > 1000, > 10 000 and > 10 000 nM at Y5, Y1, Y2 and Y4 receptors respectively) that displays no affinity for a wide range of other receptors. Does not produce a conditioned taste aversion, suppress sodium appetite or cause pica in rats. Significantly inhibits NPY-induced feeding but not through blockade of Y5 receptors.
Specifications

Chemical Identifiers

263849-50-9
528.503
1-benzoyl-2-trans-4-2-nitro-4-trifluoromethyl phenyl sulfonyl amino methyl cyclohexyl carbonyl hydrazine
N-[[4-(benzamidocarbamoyl)cyclohexyl]methyl]-2-nitro-4-(trifluoromethyl)benzenesulfonamide
C22H23F3N4O6S
BRMJFKLHWFZWOQ-UHFFFAOYSA-N
10029880
C1CC(CCC1CNS(=O)(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-])C(=O)NNC(=O)C3=CC=CC=C3

Specifications

50mg
263849-50-9
1-benzoyl-2-trans-4-2-nitro-4-trifluoromethyl phenyl sulfonyl amino methyl cyclohexyl carbonyl hydrazine
C1CC(CCC1CNS(=O)(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-])C(=O)NNC(=O)C3=CC=CC=C3
528.503
528.5
S 25585
C22H23F3N4O6S
BRMJFKLHWFZWOQ-UHFFFAOYSA-N
N-[[4-(benzamidocarbamoyl)cyclohexyl]methyl]-2-nitro-4-(trifluoromethyl)benzenesulfonamide
10029880
>98%
Videos
Documents