missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-[(tert-Butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid, Thermo Scientific™

Catalog Number p-4391229
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
65671-54-7
C12H24N2O4
260.334
HBCLYVIARHJTTL-VIFPVBQESA-N
s-2-tert-butoxycarbonyl amino-5-dimethylamino pentanoic acid, 2s-2-tert-butoxycarbonyl amino-5-dimethylamino pentanoic acid, s-2-boc-amino-5-dimethylamino pentanoic acid, s-2-tert-butoxycarbonyl amino-5-dimethylami, n∼2∼-tert-butoxycarbonyl-n∼5∼,n∼5∼-dimethyl-l-ornithine, s-2-tert-butoxycarbonylamino-5-dimethylamino pentanoic acid, l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5,n5-dimethyl, 2s-5-dimethylamino-2-tert-butoxy carbonylamino pentanoic acid
16105709
(2S)-5-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)(C)OC(=O)NC(CCCN(C)C)C(=O)O
This item is not returnable. View return policy
65671-54-7
C12H24N2O4
260.334
HBCLYVIARHJTTL-VIFPVBQESA-N
s-2-tert-butoxycarbonyl amino-5-dimethylamino pentanoic acid, 2s-2-tert-butoxycarbonyl amino-5-dimethylamino pentanoic acid, s-2-boc-amino-5-dimethylamino pentanoic acid, s-2-tert-butoxycarbonyl amino-5-dimethylami, n∼2∼-tert-butoxycarbonyl-n∼5∼,n∼5∼-dimethyl-l-ornithine, s-2-tert-butoxycarbonylamino-5-dimethylamino pentanoic acid, l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5,n5-dimethyl, 2s-5-dimethylamino-2-tert-butoxy carbonylamino pentanoic acid
16105709
(2S)-5-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)(C)OC(=O)NC(CCCN(C)C)C(=O)O
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 260.33
Percent Purity 97%
Chemical Name or Material (S)-2-[(tert-Butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'