missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylpropionic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-183248
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
7782-24-3
C9H10O2
150.18
MFCD00063139
YPGCWEMNNLXISK-ZETCQYMHSA-N
s-+-2-phenylpropionic acid, s-2-phenylpropanoic acid, 2s-2-phenylpropanoic acid, s-hydratropic acid, +-hydratropic acid, s-2-phenyl-propionic acid, s-alpha-methylbenzeneacetic acid, s-2-phenylpropionic acid, s-+-hydratropic acid, unii-988109nfbc
2724622
CHEBI:48527
(2S)-2-phenylpropanoic acid
CC(C1=CC=CC=C1)C(=O)O
This item is not returnable. View return policy
7782-24-3
C9H10O2
150.18
MFCD00063139
YPGCWEMNNLXISK-ZETCQYMHSA-N
s-+-2-phenylpropionic acid, s-2-phenylpropanoic acid, 2s-2-phenylpropanoic acid, s-hydratropic acid, +-hydratropic acid, s-2-phenyl-propionic acid, s-alpha-methylbenzeneacetic acid, s-2-phenylpropionic acid, s-+-hydratropic acid, unii-988109nfbc
2724622
CHEBI:48527
(2S)-2-phenylpropanoic acid
CC(C1=CC=CC=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 29°C to 32°C
Color White
Density 1.1000g/mL
Boiling Point 115°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3CH(C6H5)CO2H
Refractive Index 1.5223
Quantity 250 mg
Beilstein 09, 347
Specific Gravity 1.1
Specific Rotation Condition +75° (20°C c=3,CHCl3)
Specific Rotation + 75
Formula Weight 150.18
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material (S)-(+)-2-Phenylpropionic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-2-Phenylpropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'