missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methylbutyric acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3779049
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1730-91-2
C5H10O2
102.13
MFCD00063166
WLAMNBDJUVNPJU-BYPYZUCNSA-N
s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid
448893
CHEBI:38655
(2S)-2-methylbutanoic acid
CCC(C)C(=O)O
This item is not returnable. View return policy
1730-91-2
C5H10O2
102.13
MFCD00063166
WLAMNBDJUVNPJU-BYPYZUCNSA-N
s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid
448893
CHEBI:38655
(2S)-2-methylbutanoic acid
CCC(C)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9380g/mL
Boiling Point 78.0°C (15.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)COOH
Refractive Index 1.4041 to 1.4061
Quantity 1 g
Beilstein 02, 304
Specific Gravity 0.938
Specific Rotation Condition +19° (20°C c=neat)
Specific Rotation + 19.00
Formula Weight 102.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Methylbutyric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'