missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Methoxypropionic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10515301
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
23953-00-6
C4H8O3
104.11
MFCD01632589
ICPWFHKNYYRBSZ-VKHMYHEASA-N
s-2-methoxypropanoic acid, s---2-methoxypropionic acid, 2s-2-methoxypropanoic acid, propanoic acid, 2-methoxy-, 2s, unii-74k293ocx6, 2-methoxypropanoic acid, s, propanoic acid,2-methoxy-, 2s, propanoic acid, 2-methoxy-, s, propionic acid, 2-methoxy-, s--, s-2-methoxypropionic acid
5702566
(2S)-2-methoxypropanoic acid
CC(C(=O)O)OC
This item is not returnable. View return policy
23953-00-6
C4H8O3
104.11
MFCD01632589
ICPWFHKNYYRBSZ-VKHMYHEASA-N
s-2-methoxypropanoic acid, s---2-methoxypropionic acid, 2s-2-methoxypropanoic acid, propanoic acid, 2-methoxy-, 2s, unii-74k293ocx6, 2-methoxypropanoic acid, s, propanoic acid,2-methoxy-, 2s, propanoic acid, 2-methoxy-, s, propionic acid, 2-methoxy-, s--, s-2-methoxypropionic acid
5702566
(2S)-2-methoxypropanoic acid
CC(C(=O)O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 99.0°C (20.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OCH3)COOH
Refractive Index 1.4132 to 1.4152
Quantity 2.5 g
Beilstein 03, III, 444
Specific Rotation Condition −75.00 (20.00°C c=neat)
Specific Rotation − 75.00
Formula Weight 104.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Methoxypropionic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'