missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific™

Catalog Number p-150166
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
213270-36-1
C9H16O4
188.22
MFCD01091033
AJNNFMRCMKGYHA-ZETCQYMHSA-N
s-3-methoxycarbonyl-5-methylhexanoic acid, s---2-isobutylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-5-methylhexanoic acid, s-2-isobutylsuccinic acid 1-methyl ester, s-2-2-methylpropyl butanedioic acid 1-methyl ester, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2s
2734513
(3S)-3-methoxycarbonyl-5-methylhexanoic acid
CC(C)CC(CC(=O)O)C(=O)OC
This item is not returnable. View return policy
213270-36-1
C9H16O4
188.22
MFCD01091033
AJNNFMRCMKGYHA-ZETCQYMHSA-N
s-3-methoxycarbonyl-5-methylhexanoic acid, s---2-isobutylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-5-methylhexanoic acid, s-2-isobutylsuccinic acid 1-methyl ester, s-2-2-methylpropyl butanedioic acid 1-methyl ester, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2s
2734513
(3S)-3-methoxycarbonyl-5-methylhexanoic acid
CC(C)CC(CC(=O)O)C(=O)OC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Packaging Glass Bottle
Quantity 250 mg
Formula Weight 188.22
Percent Purity 95%,(98% ee)
Chemical Name or Material (S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
compliance-icons
Product Identifier
  • (S)-2-Isobutylsuccinic acid-1-methyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'