missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Hydroxy-3-buten-1-yl p-tosylate, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
Melting Point | 57.0°C to 63.0°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 1 g |
Specific Rotation Condition | − 78.50 (20.00°C c=1.13 MeOH) |
Specific Rotation | − 78.50 |
Formula Weight | 242.3 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'